MilliporeSigma
  • Valosin-containing protein (VCP/p97) plays a role in the replication of West Nile virus.
Valosin-containing protein (VCP/p97) plays a role in the replication of West Nile virus.
Virus research (2016-12-05)
Wallaya Phongphaew, Shintaro Kobayashi, Michihito Sasaki, Michael Carr, William W Hall, Yasuko Orba, Hirofumi Sawa
ABSTRACT
Valosin-containing protein (VCP) is classified as a member of the type II AAA
Sigma-Aldrich
MISSION® esiRNA, targeting human VCP
Pricing and availability is not currently available.