MilliporeSigma
  • Small RNA interactome of pathogenic E. coli revealed through crosslinking of RNase E.
Small RNA interactome of pathogenic E. coli revealed through crosslinking of RNase E.
The EMBO journal (2016-11-12)
Shafagh A Waters, Sean P McAteer, Grzegorz Kudla, Ignatius Pang, Nandan P Deshpande, Timothy G Amos, Kai Wen Leong, Marc R Wilkins, Richard Strugnell, David L Gally, David Tollervey, Jai J Tree
ABSTRACT
RNA sequencing studies have identified hundreds of non-coding RNAs in bacteria, including regulatory small RNA (sRNA). However, our understanding of sRNA function has lagged behind their identification due to a lack of tools for the high-throughput analysis of RNA-RNA interactions in bacteria. Here we demonstrate that in vivo sRNA-mRNA duplexes can be recovered using UV-crosslinking, ligation and sequencing of hybrids (CLASH). Many sRNAs recruit the endoribonuclease, RNase E, to facilitate processing of mRNAs. We were able to recover base-paired sRNA-mRNA duplexes in association with RNase E, allowing proximity-dependent ligation and sequencing of cognate sRNA-mRNA pairs as chimeric reads. We verified that this approach captures bona fide sRNA-mRNA interactions. Clustering analyses identified novel sRNA seed regions and sets of potentially co-regulated target mRNAs. We identified multiple mRNA targets for the pathotype-specific sRNA Esr41, which was shown to regulate colicin sensitivity and iron transport in E. coli Numerous sRNA interactions were also identified with non-coding RNAs, including sRNAs and tRNAs, demonstrating the high complexity of the sRNA interactome.
Sigma-Aldrich
Minimum Essential Medium Eagle, HEPES Modification, with Earle′s salts, 25 mM HEPES and sodium bicarbonate, without L-glutamine, liquid, sterile-filtered, suitable for cell culture
Pricing and availability is not currently available.