MilliporeSigma
  • Biological Characterization of a Stable Effector Functionless (SEFL) Monoclonal Antibody Scaffold in Vitro.
Biological Characterization of a Stable Effector Functionless (SEFL) Monoclonal Antibody Scaffold in Vitro.
The Journal of biological chemistry (2016-12-21)
Ling Liu, Frederick W Jacobsen, Nancy Everds, Yao Zhuang, Yan Bin Yu, Nianyu Li, Darcey Clark, Mai Phuong Nguyen, Madeline Fort, Padma Narayanan, Kei Kim, Riki Stevenson, Linda Narhi, Kannan Gunasekaran, Jeanine L Bussiere
ABSTRACT
The stable effector functionLess (SEFL) antibody was designed as an IgG1 antibody with a constant region that lacks the ability to interact with Fcγ receptors. The engineering and stability and pharmacokinetic assessments of the SEFL scaffold is described in the accompanying article (Jacobsen, F. W., Stevenson, R., Li, C., Salimi-Moosavi, H., Liu, L., Wen, J., Luo, Q., Daris, K., Buck, L., Miller, S., Ho, S-Y., Wang, W., Chen, Q., Walker, K., Wypych, J., Narhi, L., and Gunasekaran, K. (2017) J. Biol. Chem 292). The biological properties of these SEFL antibodies were assessed in a variety of human and cynomolgus monkey in vitro assays. Binding of parent molecules and their SEFL variants to human and cynomolgus monkey FcγRs were evaluated using flow cytometry-based binding assays. The SEFL variants tested showed decreased binding affinity to human and cynomolgus FcγRs compared with the wild-type IgG1 antibody. In addition, SEFL variants demonstrated no antibody-dependent cell-mediated cytotoxicity in vitro against Daudi cells with cynomolgus monkey peripheral blood mononuclear cells, and had minimal complement-dependent cytotoxicity activity similar to that of the negative control IgG2 in a CD20
Sigma-Aldrich
Formalin solution, neutral buffered, 10%, case of 48 × 15 mL, histological tissue fixative
Pricing and availability is not currently available.
Sigma-Aldrich
Monoclonal Anti-Kappa light chains-FITC antibody produced in mouse, clone A8B5, purified immunoglobulin, buffered aqueous solution
Pricing and availability is not currently available.