MilliporeSigma
  • Fluorescent nanodiamonds enable quantitative tracking of human mesenchymal stem cells in miniature pigs.
Fluorescent nanodiamonds enable quantitative tracking of human mesenchymal stem cells in miniature pigs.
Scientific reports (2017-03-31)
Long-Jyun Su, Meng-Shiue Wu, Yuen Yung Hui, Be-Ming Chang, Lei Pan, Pei-Chen Hsu, Yit-Tsong Chen, Hong-Nerng Ho, Yen-Hua Huang, Thai-Yen Ling, Hsao-Hsun Hsu, Huan-Cheng Chang
ABSTRACT
Cell therapy is a promising strategy for the treatment of human diseases. While the first use of cells for therapeutic purposes can be traced to the 19th century, there has been a lack of general and reliable methods to study the biodistribution and associated pharmacokinetics of transplanted cells in various animal models for preclinical evaluation. Here, we present a new platform using albumin-conjugated fluorescent nanodiamonds (FNDs) as biocompatible and photostable labels for quantitative tracking of human placenta choriodecidual membrane-derived mesenchymal stem cells (pcMSCs) in miniature pigs by magnetic modulation. With this background-free detection technique and time-gated fluorescence imaging, we have been able to precisely determine the numbers as well as positions of the transplanted FND-labeled pcMSCs in organs and tissues of the miniature pigs after intravenous administration. The method is applicable to single-cell imaging and quantitative tracking of human stem/progenitor cells in rodents and other animal models as well.
Sigma-Aldrich
(Tyr[SO3H]27)Cholecystokinin fragment 26-33 Amide, ≥97% (HPLC), powder
Pricing and availability is not currently available.
Sigma-Aldrich
Mesenchymal Adipogenesis Kit, Mesenchymal Stem Cell Adipogenesis Kit typically obtains 30% mature adipocytes from the rat bone marrow derived mesenchymal stem cells.
Pricing and availability is not currently available.
Sigma-Aldrich
Anti-Nuclei Antibody, clone 235-1, clone 235-1, Chemicon®, from mouse
Pricing and availability is not currently available.