MilliporeSigma
  • OR2M3: A Highly Specific and Narrowly Tuned Human Odorant Receptor for the Sensitive Detection of Onion Key Food Odorant 3-Mercapto-2-methylpentan-1-ol.
OR2M3: A Highly Specific and Narrowly Tuned Human Odorant Receptor for the Sensitive Detection of Onion Key Food Odorant 3-Mercapto-2-methylpentan-1-ol.
Chemical senses (2016-12-06)
Franziska Noe, Johannes Polster, Christiane Geithe, Matthias Kotthoff, Peter Schieberle, Dietmar Krautwurst
ABSTRACT
The detection of key food odorants appears to be an important capability of odorant receptors. Here, thiols occupy an outstanding position among the 230 known key food odorants because of their very low odor thresholds. Members of the homologous series of 3-mercapto-2-methylalkan-1-ols have been described as onion key food odorants or food constituents and are detected at logarithmically different thresholds. 3-Mercapto-2-methylpentan-1-ol being the only key food odorant within this series also has the lowest odor threshold. Most odorants typically activate combinations of odorant receptors, which may be narrowly or broadly tuned. Consequently, a specific receptor activation pattern will define an odor quality. In contrast, here we show that just 1 of the 391 human odorant receptors, OR2M3, responded exclusively to 3-mercapto-2-methylpentan-1-ol of the 190 key food odorants tested, with a half maximal effective concentration at submicromolar concentration. Moreover, neither the Denisovan OR2M3 nor the closest OR2M3 homologs from five species did respond to this compound. This outstanding specificity of extremely narrowly tuned human OR2M3 can explain both odor qualities and odor threshold trend within a homologous series of 3-mercapto-2-methylalkan-1-ols and suggests a modern human-specific, food-related function of OR2M3 in detecting a single onion key food odorant.
Sigma-Aldrich
Lipopolysaccharides from Escherichia coli K-235, purified by phenol extraction
Pricing and availability is not currently available.
Sigma-Aldrich
MEM Non-essential Amino Acid Solution (100×), without L-glutamine, liquid, sterile-filtered, BioReagent, suitable for cell culture
Pricing and availability is not currently available.