MilliporeSigma
  • Quantification of copper content with laser induced breakdown spectroscopy as a potential indicator of offal adulteration in beef.
Quantification of copper content with laser induced breakdown spectroscopy as a potential indicator of offal adulteration in beef.
Talanta (2017-04-17)
Maria P Casado-Gavalda, Yash Dixit, David Geulen, Raquel Cama-Moncunill, Xavier Cama-Moncunill, Maria Markiewicz-Keszycka, Patrick J Cullen, Carl Sullivan
ABSTRACT
Laser induced breakdown spectroscopy (LIBS) is an emerging technique in the field of food analysis which provides various advantages such as minimal sample preparation, chemical free, rapid detection, provision of spatial information and portability. In this study, LIBS was employed for quantitative analysis of copper content in minced beef samples spiked with beef liver over three independent batches. Copper content was determined with graphite furnace atomic absorption spectroscopy (GFAAS) in order to obtain reference values for modelling. Partial least square regression (PLSR) was performed to build a calibration and validation model. A calibration model with a high R
Product Description
Supelco
Copper Standard for ICP, TraceCERT®, 1 g/L Cu in nitric acid (nominal concentration)
Pricing and availability is not currently available.