MilliporeSigma
  • Detection of Reduced Susceptibility to Chlorfenapyr- and Bifenthrin-Containing Products in Field Populations of the Bed Bug (Hemiptera: Cimicidae).
Detection of Reduced Susceptibility to Chlorfenapyr- and Bifenthrin-Containing Products in Field Populations of the Bed Bug (Hemiptera: Cimicidae).
Journal of economic entomology (2017-04-12)
Aaron R Ashbrook, Michael E Scharf, Gary W Bennett, Ameya D Gondhalekar
ABSTRACT
Insecticide resistance is a major impediment for effective control of Cimex lectularius L. Previous resistance detection studies with bed bugs have focused on certain pyrethroid, neonicotinoid, organochlorine, organophosphate, and carbamate insecticides. Within the pyrethroid class, resistance studies have mostly been limited to deltamethrin, lambda-cyhalothrin, and alpha- and beta-cyfluthrin. The goal of this study was to develop diagnostic concentration bioassays for assessing bed bug susceptibility levels to chlorfenapyr- and bifenthrin-containing products. First, glass vial and filter paper bioassay methods were compared for their utility in susceptibility monitoring. Statistical comparison of toxicity data between bioassays indicated that the vial assay was less confounded by assay susbtrate effects, required less insecticide, and was faster, especially for chlorfenapyr. Next, using vial diagnostic concentrations (LC99) for each insecticide, 10 laboratory-adapted field strains and the Harlan lab-susceptible strain were screened for susceptibility to chlorfenapyr and bifenthrin. The results of this study reveal recent bed bug susceptibility levels to certain chlorfenapyr- and bifenthrin-containing products. Reduced susceptibility was detected in three and five field strains to chlorfenapyr and bifenthrin, respectively. Detection of reduced susceptibility suggests that certain strains may be segregating toward greater chlorfenapyr and bifenthrin resistance. These results merit continuous resistance monitoring efforts to detect chlorfenapyr and bifenthrin susceptibility shifts. Additionally, to reduce insecticide selection pressures and delay resistance development, adoption of integrated bed bug control strategies that combine chemical and nonchemical methods is recommended.
Product Description
Supelco
Chlorfenapyr, PESTANAL®, analytical standard
Pricing and availability is not currently available.