- Gene Expression Profiling of Tuberculous Meningitis Co-infected with HIV.
Gene Expression Profiling of Tuberculous Meningitis Co-infected with HIV.
Journal of proteomics & bioinformatics (2012-09-01)
Ghantasala S Sameer Kumar, Abhilash K Venugopal, Manoj Kumar Kashyap, Rajesh Raju, Arivusudar Marimuthu, Shyam Mohan Palapetta, Yashwanth Subbanayya, Renu Goel, Ankit Chawla, Jyoti Bajpai Dikshit, Pramila Tata, H C Harsha, Jagadeesha Maharudraiah, Y L Ramachandra, Parthasarathy Satishchandra, T S Keshava Prasad, Akhilesh Pandey, Anita Mahadevan, S K Shankar
PMID27053842
ABSTRACT
Tuberculous meningitis (TBM) is a fatal form of
Sigma-Aldrich
Anti-TYMP antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
Pricing and availability is not currently available.