MilliporeSigma
  • Taurolidine/Citrate Lock Therapy for Primary Prevention of Catheter-Related Infections in Cancer Patients: Results of a Prospective, Randomized, Phase IV Trial (ATAPAC).
Taurolidine/Citrate Lock Therapy for Primary Prevention of Catheter-Related Infections in Cancer Patients: Results of a Prospective, Randomized, Phase IV Trial (ATAPAC).
Oncology (2017-05-04)
Raffaele Longo, Mathieu Llorens, Christophe Goetz, Christian Platini, Nada Eid, Jocelyne Sellies, Nadia Ouamara, Philippe Quétin
ABSTRACT
Totally implantable venous access port (TIVAP)-related infections (RIs) remain a serious health problem in cancer patients receiving an intravenous (i.v.) therapy. The ATAPAC study was a prospective, randomized, monocentric, phase IV trial evaluating the efficacy of taurolidine lock solution versus standard saline solution for primary TIVAP-RI prevention in nonhematological cancer patients receiving i.v. chemotherapy. The primary endpoint was the TIVAP-RI incidence rate. From December 2014 to September 2015, 163 patients were enrolled in the study (taurolidine: n = 86 vs. n = 77). Four patients in the control group (5%) had a Staphylococcus epidermidis TIVAP-RI, and 1 patient (1%) in the taurolidine group had a Staphylococcus aureus infection. The TIVAP-RI incidence rate was 0.4 and 0.1‰ catheter-days, respectively (p = 0.21). The infection-free TIVAP survival was not statistically significant (p = 0.09). TIVAP-RI required a total of 22 hospitalization days in the taurolidine group versus 106 days in the control arm with associated costs of EUR 4,849 and EUR 36,020, respectively. Taurolidine-related toxicity was transitory and classified as grade I. The ATAPAC trial did not show a significant risk-infection reduction by TauroLock™. A larger, prospective, randomized trial is needed to assess TauroLock efficacy for primary TIVAP-RI prevention in low-risk cancer patients.
Sigma-Aldrich
Taurolidine, >97% (NMR), powder
Pricing and availability is not currently available.