MilliporeSigma
  • Insulin-like growth factor 1 receptor regulates hypothermia during calorie restriction.
Insulin-like growth factor 1 receptor regulates hypothermia during calorie restriction.
Proceedings of the National Academy of Sciences of the United States of America (2017-08-23)
Rigo Cintron-Colon, Manuel Sanchez-Alavez, William Nguyen, Simone Mori, Ruben Gonzalez-Rivera, Tiffany Lien, Tamas Bartfai, Saba Aïd, Jean-Christophe François, Martin Holzenberger, Bruno Conti
ABSTRACT
When food resources are scarce, endothermic animals can lower core body temperature (T
Sigma-Aldrich
Picropodophyllotoxin, ≥96% (HPLC), powder
Pricing and availability is not currently available.