MilliporeSigma
  • Improved Self-cleaning Properties of an Efficient and Easy to Scale up TiO
Improved Self-cleaning Properties of an Efficient and Easy to Scale up TiO
Scientific reports (2017-08-27)
Rima J Isaifan, Ayman Samara, Wafa Suwaileh, Daniel Johnson, Wubulikasimu Yiming, Amir A Abdallah, Brahim Aïssa
ABSTRACT
Transparent titania coatings have self-cleaning and anti-reflection properties (AR) that are of great importance to minimize soiling effect on photovoltaic modules. In this work, TiO
Sigma-Aldrich
Tetraethylene glycol, 99%
Pricing and availability is not currently available.