MilliporeSigma
Preparation and Self-Assembly of Dendronized Janus Fe
ACS nano (2017-08-05)
Davit Jishkariani, Yaoting Wu, Da Wang, Yang Liu, Alfons van Blaaderen, Christopher B Murray
ABSTRACT
Janus nanoparticles (NPs) often referred to as nanosized analogs of molecular surfactants are amphiphilic structures with potential applications in materials science, biomedicine, and catalysis, and their synthesis and self-assembly into complex architectures remain challenging. Here, we demonstrate the preparation of Janus heterodimers via asymmetric functionalization of Fe
Sigma-Aldrich
1-Octadecene, technical grade, 90%
Pricing and availability is not currently available.
Sigma-Aldrich
2,2-Dimethoxypropane, reagent grade, 98%
Pricing and availability is not currently available.