MilliporeSigma
  • Chromodomain Protein CDYL Acts as a Crotonyl-CoA Hydratase to Regulate Histone Crotonylation and Spermatogenesis.
Chromodomain Protein CDYL Acts as a Crotonyl-CoA Hydratase to Regulate Histone Crotonylation and Spermatogenesis.
Molecular cell (2017-08-15)
Shumeng Liu, Huajing Yu, Yongqing Liu, Xinhua Liu, Yu Zhang, Chen Bu, Shuai Yuan, Zhe Chen, Guojia Xie, Wanjin Li, Bosen Xu, Jianguo Yang, Lin He, Tong Jin, Yundong Xiong, Luyang Sun, Xiaohui Liu, Chunsheng Han, Zhongyi Cheng, Jing Liang, Yongfeng Shang
ABSTRACT
Lysine crotonylation (Kcr) is a newly identified histone modification that is associated with active transcription in mammalian cells. Here we report that the chromodomain Y-like transcription corepressor CDYL negatively regulates histone Kcr by acting as a crotonyl-CoA hydratase to convert crotonyl-CoA to β-hydroxybutyryl-CoA. We showed that the negative regulation of histone Kcr by CDYL is intrinsically linked to its transcription repression activity and functionally implemented in the reactivation of sex chromosome-linked genes in round spermatids and genome-wide histone replacement in elongating spermatids. Significantly, Cdyl transgenic mice manifest dysregulation of histone Kcr and reduction of male fertility with a decreased epididymal sperm count and sperm cell motility. Our study uncovers a biochemical pathway in the regulation of histone Kcr and implicates CDYL-regulated histone Kcr in spermatogenesis, adding to the understanding of the physiology of male reproduction and the mechanism of the spermatogenic failure in AZFc (Azoospermia Factor c)-deleted infertile men.
Sigma-Aldrich
Nuclei Isolation Kit: Nuclei PURE Prep, sufficient for 15 nuclei preparations (~1-10×107 cells or 1g of tissue per preparation)
Pricing and availability is not currently available.
Sigma-Aldrich
Anti-trimethyl Histone H3 (Lys27) Antibody, Trial Size, from rabbit, purified by affinity chromatography
Pricing and availability is not currently available.