MilliporeSigma
Bradykinin B2 and dopamine D2 receptors form a functional dimer.
Biochimica et biophysica acta (2017-08-02)
Anna Niewiarowska-Sendo, Agnieszka Polit, Monika Piwowar, Magdalena Tworzydło, Andrzej Kozik, Ibeth Guevara-Lora
ABSTRACT
In recent years a wide range of studies have shown that G protein-coupled receptors modulate a variety of cell functions through the formation of dimers. For instance, there is growing evidence for the dimerization of bradykinin or dopamine receptors, both as homodimers and heterodimers. A discovery of direct interactions of angiotensin II receptors with bradykinin 2 receptor (B2R) or dopamine D2 (D2R) receptor has led to a hypothesis on a potential dimerization between two latter receptors. In this study, we have demonstrated a constitutive colocalization of receptors on the membranes of HEK293 cells transiently transfected with plasmid vectors encoding B2R and D2R, fused with fluorescent proteins. The receptor colocalization was significantly enhanced by specific agonists of B2R or D2R after 5min following the addition, whereas simultaneous stimulation with these agonists did not influence the B2R/D2R colocalization level. In addition, B2R-D2R heterodimerization was confirmed with FLIM-FRET technique. The most characteristic signaling pathways for B2R and D2R, dependent on intracellular Ca
Product Description
Supelco
Bacitracin A, VETRANAL®, analytical standard, main stereoisomer ~50 %
Pricing and availability is not currently available.
Sigma-Aldrich
Sumanirole maleate, ≥98% (HPLC)
Pricing and availability is not currently available.