MilliporeSigma
  • Etiolated Stem Branching Is a Result of Systemic Signaling Associated with Sucrose Level.
Etiolated Stem Branching Is a Result of Systemic Signaling Associated with Sucrose Level.
Plant physiology (2017-09-02)
Bolaji Babajide Salam, Siva Kumar Malka, Xiaobiao Zhu, Huiling Gong, Carmit Ziv, Paula Teper-Bamnolker, Naomi Ori, Jiming Jiang, Dani Eshel
ABSTRACT
The potato (
Sigma-Aldrich
β-Gentiobiose, ≥85% (remainder primarily α-anomer)
Pricing and availability is not currently available.