MilliporeSigma
  • Crocus sativus, Serenoa repens and Pinus massoniana extracts modulate inflammatory response in isolated rat prostate challenged with LPS.
Crocus sativus, Serenoa repens and Pinus massoniana extracts modulate inflammatory response in isolated rat prostate challenged with LPS.
Journal of biological regulators and homeostatic agents (2017-09-12)
A Chiavaroli, L Recinella, C Ferrante, M Locatelli, S Carradori, N Macchione, G Zengin, L Leporini, S Leone, S Martinotti, L Brunetti, M Vacca, L Menghini, G Orlando
ABSTRACT
Prostatitis is a common prostate disease that could be promoted by bacterial or non-bacterial infectious agents. In addition, inflammatory pathways involved in prostatitis have been increasingly studied, and herbal extracts endowed with anti-inflammatory effects are under investigation, individually or in combination, for their efficacy in alleviating the burden of inflammation, with possible improvements in symptoms. Serenoa repens (Serenoa), in combination with Crocus sativus (Crocus) and Pinus massoniana (Pinus), has previously shown to improve sexual function and limit urinary symptoms in patients suffering from concomitant erectile dysfunction and lower urinary tract symptoms. In this context, the aim of the present study is to evaluate the efficacy of Serenoa, Crocus and Pinus extracts, either alone or in combination, on immortalized prostate cells (PC3) and in an experimental model of bacterial prostatitis constituted by ex vivo prostate specimens challenged with lipopolysaccharide (LPS). We found that the tested extracts were able to reduce ROS production by PC3 cells and NFkB and PGE2 activity in prostate specimens challenged with LPS. In addition, the pharmacological association of the extracts displayed synergistic effects indicating a rational use of the mixture of the tested extracts as a novel anti-oxidant and anti-inflammatory formulation in bacterial prostatitis. Finally, we performed analytical and in vitro evaluation to better characterize the phytochemical profile and the mechanism of action of selected secondary metabolites.
Sigma-Aldrich
2-Hydroxycinnamic acid, predominantly trans, 97%
Pricing and availability is not currently available.