MilliporeSigma
  • LC-MS/MS quantification of 7α-hydroxy-4-cholesten-3-one (C4) in rat and monkey plasma.
LC-MS/MS quantification of 7α-hydroxy-4-cholesten-3-one (C4) in rat and monkey plasma.
Journal of chromatography. B, Analytical technologies in the biomedical and life sciences (2017-09-16)
Lijuan Kang, Thomas M Connolly, Naidong Weng, Wenying Jian
ABSTRACT
7α-hydroxy-4-cholesten-3-one (C4) is an oxidative enzymatic product of cholesterol metabolism via cholesterol 7α-hydroxylase, an enzyme also known as cholesterol 7-alpha-monooxygenase or cytochrome P450 7A1 (CYP7A1). C4 is a stable intermediate in the rate limiting pathway of bile acid biosynthesis. Previous studies showed that plasma C4 levels correlated with CYP7A1 enzymatic activity and could serve as a biomarker for bile acid synthesis. Here we developed and qualified a simple and robust high-throughput method using liquid chromatography coupled with tandem mass spectrometry (LC-MS/MS) to quantify C4 in rat and monkey plasma. As C4 being an endogenous compound, this method used calibration standards in 50/50: acetonitrile/water (v/v). In order to mimic the incurred samples, quality control samples were prepared in the authentic plasma. Stable isotope labeled C4 (C4-d
Product Description
Supelco
7α-Hydroxy-4-cholesten-3-one, ≥95.0% (HPLC)
Pricing and availability is not currently available.