MilliporeSigma
  • A Bifunctional MAPK/PI3K Antagonist for Inhibition of Tumor Growth and Metastasis.
A Bifunctional MAPK/PI3K Antagonist for Inhibition of Tumor Growth and Metastasis.
Molecular cancer therapeutics (2017-08-05)
Stefanie Galbán, April A Apfelbaum, Carlos Espinoza, Kevin Heist, Henry Haley, Karan Bedi, Mats Ljungman, Craig J Galbán, Gary D Luker, Marcian Van Dort, Brian D Ross
ABSTRACT
Responses to targeted therapies frequently are brief, with patients relapsing with drug-resistant tumors. For oncogenic MEK and BRAF inhibition, drug resistance commonly occurs through activation of PI3K/AKT/mTOR signaling and immune checkpoint modulation, providing a robust molecular target for concomitant therapy. Here, we evaluated the efficacy of a bifunctional kinase inhibitor (ST-162) that concurrently targets MAPK and PI3K signaling pathways. Treatment with ST-162 produced regression of mutant KRAS- or BRAF-addicted xenograft models of colorectal cancer and melanoma and stasis of BRAF/PTEN-mutant melanomas. Combining ST-162 with immune checkpoint blockers further increased efficacy in a syngeneic KRAS-mutant colorectal cancer model. Nascent transcriptome analysis revealed a unique gene set regulated by ST-162 related to melanoma metastasis. Subsequent mouse studies revealed ST-162 was a potent inhibitor of melanoma metastasis to the liver. These findings highlight the significant potential of a single molecule with multikinase activity to achieve tumor control, overcome resistance, and prevent metastases through modulation of interconnected cell signaling pathways.
Sigma-Aldrich
MEK1, active, GST tagged human, PRECISIO® Kinase, recombinant, expressed in baculovirus infected Sf9 cells, ≥70% (SDS-PAGE), buffered aqueous glycerol solution
Pricing and availability is not currently available.