MilliporeSigma
  • Iron absorption from oral iron supplements given on consecutive versus alternate days and as single morning doses versus twice-daily split dosing in iron-depleted women: two open-label, randomised controlled trials.
Iron absorption from oral iron supplements given on consecutive versus alternate days and as single morning doses versus twice-daily split dosing in iron-depleted women: two open-label, randomised controlled trials.
The Lancet. Haematology (2017-10-17)
Nicole U Stoffel, Colin I Cercamondi, Gary Brittenham, Christophe Zeder, Anneke J Geurts-Moespot, Dorine W Swinkels, Diego Moretti, Michael B Zimmermann
ABSTRACT
Current guidelines to treat iron deficiency recommend daily provision of ferrous iron divided through the day to increase absorption. However, daily dosing and split dosing might increase serum hepcidin and decrease iron absorption from subsequent doses. Our study aim was to compare iron absorption from oral iron supplements given on consecutive versus alternate days and given as single morning doses versus twice-daily split dosing. We did two prospective, open-label, randomised controlled trials assessing iron absorption using ( For study 1, 40 women were enrolled on Oct 15-29, 2015. 21 women were assigned to the consecutive-day group and 19 to the alternate-day group. At the end of treatment (14 days for the consecutive-day group and 28 days for the alternate-day group), geometric mean (-SD, +SD) cumulative fractional iron absorptions were 16·3% (9·3, 28·8) in the consecutive-day group versus 21·8% (13·7, 34·6) in the alternate-day group (p=0·0013), and cumulative total iron absorption was 131·0 mg (71·4, 240·5) versus 175·3 mg (110·3, 278·5; p=0·0010). During the first 14 days of supplementation in both groups, serum hepcidin was higher in the consecutive-day group than the alternate-day group (p=0·0031). In study 2, 20 women were enrolled between Aug 13 and 18, 2015. Ten women were assigned to receive once-daily dosing and ten were assigned to receive twice-daily divided dosing. No significant differences were seen in fractional (day 1-3 geometric mean: 11·8% [7·1, 19·4] once daily vs 13·1% [8·2, 20·7] twice daily; p=0·33) or total iron absorption (day 1-3: 44·3 mg [29·4, 66·7] once daily vs 49·4 [35·2, 69·4] twice daily; p=0·33) between the two dosing regimens. Twice-daily divided doses resulted in a higher serum hepcidin concentration than once-daily dosing (p=0·013). No grade 3 or 4 adverse events were reported in either study. In iron-depleted women, providing iron supplements daily as divided doses increases serum hepcidin and reduces iron absorption. Providing iron supplements on alternate days and in single doses optimises iron absorption and might be a preferable dosing regimen. These findings should be confirmed in iron-deficient anaemic patients. Swiss National Science Foundation, Bern, Switzerland.
Sigma-Aldrich
Human FABP2 / Fatty Acid-Binding Protein, Intestinal ELISA Kit, for serum, plasma, cell culture supernatants and urine
Pricing and availability is not currently available.