MilliporeSigma
  • Serum lipid profile, sleep-disordered breathing and blood pressure in the elderly: a 10-year follow-up of the PROOF-SYNAPSE cohort.
Serum lipid profile, sleep-disordered breathing and blood pressure in the elderly: a 10-year follow-up of the PROOF-SYNAPSE cohort.
Sleep medicine (2017-11-22)
Denis Monneret, Jean-Claude Barthélémy, David Hupin, Delphine Maudoux, Sébastien Celle, Emilia Sforza, Frédéric Roche
ABSTRACT
Dyslipidemia, sleep-disordered breathing (SDB) and hypertension are comorbid factors evidenced in adults, but poorly studied in old people. This study aimed to examine the long-term evolution of the serum lipid profile, and its relationships with SDB and blood pressure (BP) in the elderly. A ten-year follow-up of the prospective Prognostic Indicator of Cardiovascular and Cerebrovascular Events (PROOF) and the Autonomic Nervous System Activity, Aging and Sleep Apnea/Hypopnea (SYNAPSE) cohort, which initially included 1011 elderly subjects from the general population, and who were untreated by continuous positive airway pressure (CPAP). Serum lipid profile, respiratory polygraphy for SDB and ambulatory blood pressure monitoring were performed. A total of 266 subjects (male/female 150/116; age 66.2 ± 0.8 years) were reassessed after 9.6 ± 0.7 years (age 75.8 ± 1.2 years). The prevalence of high-risk dyslipidemia decreased from 61.3 to 44.4%, and hypertension from 57.9 to 27.4%. The nocturnal oxygen desaturation index increased (mean + 2.3 ± 6.7 events/hour; p < 0.001), while the mean SaO The results suggested that the observed worsening of nocturnal oxygen desaturation after 10 years in the elderly was independent of the change in circulating lipids, and not influenced by lipid-lowering treatments. However, the variation in blood pressure remained associated with aging, waist/hip and LDL-C/HDL-C ratios. NCT00759304 and NCT00766584 at clinicaltrials.gov.
Sigma-Aldrich
Pyrvinium pamoate salt hydrate, ≥98% (HPLC)
Pricing and availability is not currently available.