- Enhanced mobility CsPbI
Enhanced mobility CsPbI
Science advances (2017-11-04)
Erin M Sanehira, Ashley R Marshall, Jeffrey A Christians, Steven P Harvey, Peter N Ciesielski, Lance M Wheeler, Philip Schulz, Lih Y Lin, Matthew C Beard, Joseph M Luther
PMID29098184
ABSTRACT
We developed lead halide perovskite quantum dot (QD) films with tuned surface chemistry based on A-site cation halide salt (AX) treatments. QD perovskites offer colloidal synthesis and processing using industrially friendly solvents, which decouples grain growth from film deposition, and at present produce larger open-circuit voltages (