MilliporeSigma
  • Small nuclear ribonucleoprotein associated polypeptide N accelerates cell proliferation in pancreatic adenocarcinoma.
Small nuclear ribonucleoprotein associated polypeptide N accelerates cell proliferation in pancreatic adenocarcinoma.
Molecular medicine reports (2015-08-12)
Jin Ma, Zhuo Zhang, Jiancheng Wang
ABSTRACT
The spliceosome, the large RNA‑protein molecular complex, is crucial for pre‑mRNA splicing. Several antitumor drugs have been found to tightly bind to the components of the spliceosome and mutations in the spliceosome have been reported in several types of cancer. However, the involvement of the spliceosome in pancreatic adenocarcinoma remains unclear. In the present study, small nuclear ribonucleoprotein associated polypeptide N (SNRPN), a key constituent of spliceosomes, was disrupted in BxPC‑3 pancreatic adenocarcinoma cells using lentivirus‑mediated RNA interference (RNAi). It was found that knockdown of SNRPN reduced the proliferation ability of BxPC‑3 cells, as determined by an MTT assay. Furthermore, cell colony formation was impaired in SNRPN depleted adenocarcinoma cells and cell cycle analysis showed that depletion of SNRPN led to S phase cell cycle arrest and apoptosis. These results suggest that SNRPN is a key player in pancreatic adenocarcinoma cell growth, and targeted loss of SNRPN may be a potential therapeutic method for pancreatic cancer.
Sigma-Aldrich
Anti-SNRPB antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
Pricing and availability is not currently available.