MilliporeSigma
  • Increased SBPase activity improves photosynthesis and grain yield in wheat grown in greenhouse conditions.
Increased SBPase activity improves photosynthesis and grain yield in wheat grown in greenhouse conditions.
Philosophical transactions of the Royal Society of London. Series B, Biological sciences (2017-08-16)
Steven M Driever, Andrew J Simkin, Saqer Alotaibi, Stuart J Fisk, Pippa J Madgwick, Caroline A Sparks, Huw D Jones, Tracy Lawson, Martin A J Parry, Christine A Raines
ABSTRACT
To meet the growing demand for food, substantial improvements in yields are needed. This is particularly the case for wheat, where global yield has stagnated in recent years. Increasing photosynthesis has been identified as a primary target to achieve yield improvements. To increase leaf photosynthesis in wheat, the level of the Calvin-Benson cycle enzyme sedoheptulose-1,7-biphosphatase (SBPase) has been increased through transformation and expression of a
Sigma-Aldrich
Anti-Transforming Growth Factor-β, pan antibody produced in rabbit, IgG fraction of antiserum, lyophilized powder
Pricing and availability is not currently available.