MilliporeSigma
  • Determination of seven pyrethroid pesticide residues in vegetables by gas chromatography using carboxylated multi-walled carbon nanotubes as dispersion solid phase extraction sorbent.
Determination of seven pyrethroid pesticide residues in vegetables by gas chromatography using carboxylated multi-walled carbon nanotubes as dispersion solid phase extraction sorbent.
Food additives & contaminants. Part A, Chemistry, analysis, control, exposure & risk assessment (2017-09-25)
Peng Sun, Yuling Gao, Chong Xu, Yongfu Lian
ABSTRACT
A novel carboxylated multi-walled carbon nanotubes (MWCNTs-COOH) dispersive solid phase extraction (d-SPE) method combined with gas chromatography (GC) with an electron capture detector (ECD) was developed for the determination of seven pyrethroid pesticides in cucumber, spinach, eggplant, tomato and carrot. We optimised d-SPE conditions including the type and volume of the extractant, the type and amount of the sorbent, and shaking time. Under the optimal conditions, the linear range was from 2.0 to 2000 μg kg
Sigma-Aldrich
ML204, ≥98% (HPLC)
Pricing and availability is not currently available.