MilliporeSigma
  • Herrmann-Beller phosphapalladacycle-catalyzed addition of alkynes to norbornadienes.
Herrmann-Beller phosphapalladacycle-catalyzed addition of alkynes to norbornadienes.
Organic letters (2006-09-08)
Alphonse Tenaglia, Laurent Giordano, Gérard Buono
ABSTRACT
Herrmann-Beller (H-B) phosphapalladacycle selectively catalyzed the addition of terminal alkynes across one double bond of norbornadiene to afford exo-5-alkynyl-bicyclo[2.2.1]hept-2-enes. The reaction shows general applicability to various functionalized alkynes and bicyclo[2.2.1]hepta-2,5-dienes. Insights into the mechanism of this reaction are discussed.
Sigma-Aldrich
trans-Bis(acetato)bis[o-(di-o-tolylphosphino)benzyl]dipalladium(II), 98%
Pricing and availability is not currently available.