MilliporeSigma
  • Preparation of core-shell NaGdF 4 nanoparticles doped with luminescent lanthanide ions to be used as upconversion-based probes.
Preparation of core-shell NaGdF 4 nanoparticles doped with luminescent lanthanide ions to be used as upconversion-based probes.
Nature Protocols (2014)
Wang F