MilliporeSigma
  • Evaluation of monoacylglycerol lipase as a therapeutic target in a transgenic mouse model of ALS.
Evaluation of monoacylglycerol lipase as a therapeutic target in a transgenic mouse model of ALS.
Neuropharmacology (2017-04-05)
Noemi Pasquarelli, Michael Engelskirchen, Johannes Hanselmann, Sascha Endres, Christoph Porazik, Hanna Bayer, Eva Buck, Meliha Karsak, Patrick Weydt, Boris Ferger, Anke Witting
ABSTRACT
Amyotrophic lateral sclerosis (ALS) is a fatal neurodegenerative disease of the motor neuron system with limited therapeutic options. While an increasing number of ALS patients can be linked to a small number of autosomal-dominantly inherited cases, most cases are termed sporadic. Both forms are clinically and histopathologically indistinguishable, raising the prospect that they share key pathogenic steps, including potential therapeutic intervention points. The endocannabinoid system is emerging as a versatile, druggable therapeutic target in the CNS and its dysregulation is an early hallmark of neurodegeneration. Whether this is a defense mechanism or part of the pathogenesis remains to be determined. The neuroprotective and anti-inflammatory endocannabinoid 2-arachidonoylglycerol (2-AG), which is degraded by monoacylglycerol lipase (MAGL), accumulates in the spinal cords of transgenic models of ALS. We tested the hypothesis that this 2-AG increase is a protective response in the low-copy SOD1
Sigma-Aldrich
KML29, ≥98% (HPLC)
Pricing and availability is not currently available.