MilliporeSigma
  • Human target validation of phosphoinositide 3-kinase (PI3K)β: effects on platelets and insulin sensitivity, using AZD6482 a novel PI3Kβ inhibitor.
Human target validation of phosphoinositide 3-kinase (PI3K)β: effects on platelets and insulin sensitivity, using AZD6482 a novel PI3Kβ inhibitor.
Journal of thrombosis and haemostasis : JTH (2012-08-22)
S Nylander, B Kull, J A Björkman, J C Ulvinge, N Oakes, B M Emanuelsson, M Andersson, T Skärby, T Inghardt, O Fjellström, D Gustafsson
ABSTRACT
Based on in vitro and animal data, PI3Kβ is given an important role in platelet adhesion and aggregation but its role in insulin signaling is unclear. To strengthen the PI3Kβ target validation using the novel, short-acting inhibitor AZD6482. AZD6482 is a potent, selective and ATP competitive PI3Kβ inhibitor (IC(50) 0.01 μm). A maximal anti-platelet effect was achieved at 1 μm in the in vitro and ex vivo tests both in dog and in man. In dog, in vivo AZD6482 produced a complete anti-thrombotic effect without an increased bleeding time or blood loss. AZD6482 was well tolerated in healthy volunteers during a 3-h infusion. The ex vivo anti-platelet effect and minimal bleeding time prolongation in the dog model translated well to data obtained in healthy volunteers. AZD6482 inhibited insulin-induced human adipocyte glucose uptake in vitro (IC(50) of 4.4 μm). In the euglycemic hyperinsulinemic clamp model, in rats, glucose infusion rate was not affected at 2.3 μm but reduced by about 60% at a plasma exposure of 27 μm. In man, the homeostasis model analysis (HOMA) index increased by about 10-20% at the highest plasma concentration of 5.3 μm. This is the first human target validation for PI3Kβ inhibition as anti-platelet therapy showing a mild and generalized antiplatelet effect attenuating but not completely inhibiting multiple signaling pathways with an impressive separation towards primary hemostasis. AZD6482 at 'supratherapeutic' plasma concentrations may attenuate insulin signaling, most likely through PI3Kα inhibition.
Sigma-Aldrich
AZD6482, ≥98% (HPLC)
Pricing and availability is not currently available.