MilliporeSigma
Kinetic mechanism of glutaryl-CoA dehydrogenase.
Biochemistry (2006-12-21)
K Sudhindra Rao, Mark Albro, Timothy M Dwyer, Frank E Frerman
ABSTRACT
Glutaryl-CoA dehydrogenase (GCD) is a homotetrameric enzyme containing one noncovalently bound FAD per monomer that oxidatively decarboxylates glutaryl-CoA to crotonyl-CoA and CO2. GCD belongs to the family of acyl-CoA dehydrogenases that are evolutionarily conserved in their sequence, structure, and function. However, there are differences in the kinetic mechanisms among the different acyl-CoA dehydrogenases. One of the unanswered aspects is that of the rate-determining step in the steady-state turnover of GCD. In the present investigation, the major rate-determining step is identified to be the release of crotonyl-CoA product because the chemical steps and reoxidation of reduced FAD are much faster than the turnover of the wild-type GCD. Other steps are only partially rate-determining. This conclusion is based on the transit times of the individual reactions occurring in the active site of GCD.
MATERIALS
Product Number
Brand
Product Description
Sigma-Aldrich
Glutaryl coenzyme A lithium salt, ≥90%
Pricing and availability is not currently available.