MilliporeSigma
HPLC Analysis of 2-Chloroethanol Content in Hydroxyzine HCl on SeQuant® ZIC-HILIC with Refractive Index Detection
HPLC Analysis of 2-Chloroethanol Content in Hydroxyzine HCl on SeQuant® ZIC-HILIC with Refractive Index Detection application for HPLC
CONDITIONS
column
SeQuant ZIC-HILIC 150 X 4.6 mm, 5.0 uM, 200 Angstrom (1.51455.0001)
mobile phase
[A] 40mM ammonium acetate; [B] acetonitrile;; (25:75, A:B)
flow rate
0.5 mL/min
column temp.
40 °C
detector
Shimadzu Proninance, RI (Detection Range 1, Cell temp. 40 °C
injection
25 μL
sample
100 ppm
Description
Legal Information
SeQuant is a registered trademark of Merck KGaA, Darmstadt, Germany