MilliporeSigma
HPLC Analysis of Mixture of Anisole and Related Compounds on Chromolith® Performance Diol 100-4.6mm
HPLC Analysis of Mixture of Anisole and Related Compounds on Chromolith® Performance Diol 100-4.6mm application for HPLC
CONDITIONS
column
Chromolith® Performance DIOL, 100 x 4.6 mm (1.53172.0001)
mobile phase
[A] n-Heptane ; [B] 1,4-Dioxane; (95:5, A:B)
flow rate
1.3 mL/min
pressure
203 psi (14 Bar)
column temp.
23 °C
detector
254 nm
injection
5 μL
sample
Anisole 390μg/ml, 3-Nitroanisole 70μg/ml, 4-Nitroanisole 260μg/ml, 2-Nitroanisole 180μg/ml (Diluent: Mobile phase)
Description
Legal Information
Chromolith is a registered trademark of Merck KGaA, Darmstadt, Germany