MilliporeSigma
HPLC Analysis of Purines and Pyrimidines on SeQuant® ZIC-HILIC
HPLC Analysis of Purines and Pyrimidines on SeQuant® ZIC-HILIC application for HPLC
CONDITIONS
column
SeQuant ZIC-HILIC, PEEK, 150 x 2.1 mm, 5 μm, 200 Å (1.50454.0001)
mobile phase
[A] acetonitrile; [B] ammonium formate; (80:20, A:B)
flow rate
0.1 mL/min
pressure
(1.4 MPa)
detector
254 nm
injection
2 μL
Description
Legal Information
SeQuant is a registered trademark of Merck KGaA, Darmstadt, Germany