Sign In to View Organizational & Contract Pricing
Select a Size
Product AMBH93E4C670 is currently not available for purchase. Contact Technical Service
Skip To
Iupac Name
2-butylidene-3-((2'-(2-trityl-2H-tetrazol-5-yl)biphenyl-4-yl)methyl)-1,3-diazaspiro[4.4]nonan-4-one
Inchi Code
1S/C44H42N6O/c1-2-3-25-40-45-43(30-15-16-31-43)42(51)49(40)32-33-26-28-34(29-27-33)38-23-13-14-24-39(38)41-46-48-50(47-41)44(35-17-7-4-8-18-35,36-19-9-5-10-20-36)37-21-11-6-12-22-37/h4-14,17-29,45H,2-3,15-16,30-32H2,1H3
InChI key
OVWHRCBDNHCJMA-UHFFFAOYSA-N
storage temp.
Sealed in dry, store in freezer, under -20C
Purity
97%
Country of Origin
CN
Shipping Temperature
Shipped in cold pack
Physical Form
solid
Product Link
https://www.ambeed.com/products/886999-35-5.html
Signal Word
Danger
Hazard Statements
Precautionary statements
MSDS
https://www.ambeed.com/products/886999-35-5.html
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.

