Skip to Content
MilliporeSigma

ENA341292229

Enamine

(2S)-1-carbamoylpyrrolidine-2-carboxylic acid

Sign In to View Organizational & Contract Pricing

Select a Size

Pricing and availability is not currently available. Contact Technical Service

About This Item

CAS Number:
Molecular Weight:
158.16
MDL number:

Skip To

Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist

mp

203-205

Iupac Name

(2S)-1-(aminocarbonyl)-2-pyrrolidinecarboxylic acid

Inchi Code

1S/C6H10N2O3/c7-6(11)8-3-1-2-4(8)5(9)10/h4H,1-3H2,(H2,7,11)(H,9,10)/t4-/m0/s1

InChI key

RTNFBJXLYMERBB-BYPYZUCNSA-N

storage temp.

RT

Purity

95

Country of Origin

UA

Shipping Temperature

Normal

Physical Form

powder

Product Link

https://www.enaminestore.com/catalog/EN300-87536

Melting Point

203-205

pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

MSDS

https://www.enaminestore.com/catalog/EN300-87536


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service