Sign In to View Organizational & Contract Pricing.
Select a Size
50 MG
$448.50
100 MG
$669.50
250 MG
$955.50
500 MG
$1,505.40
1 G
$1,930.50
2.5 G
$3,785.60
5 G
$5,600.40
10 G
$8,305.70
Vendor SKU: EN300-7440523-50MG
$448.50
Ships in 7 days - from Aldrich Partner.
About This Item
Skip To
Iupac Name
sodium 5-cyclopropyloxazole-2-carboxylate
Inchi Code
1S/C7H7NO3.Na/c9-7(10)6-8-3-5(11-6)4-1-2-4;/h3-4H,1-2H2,(H,9,10);/q;+1/p-1
InChI key
DMKLTNKAGCFOSW-UHFFFAOYSA-M
storage temp.
RT
Salt Data
[Na+]
Purity
95
Country of Origin
UA
Shipping Temperature
Normal
Physical Form
powder
Product Link
https://www.enaminestore.com/catalog/EN300-7440523
Signal Word
Warning
Hazard Statements
MSDS
https://www.enaminestore.com/catalog/EN300-7440523
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service