Sign In to View Organizational & Contract Pricing
Select a Size
Product FLUH99C809D1 is currently not available for purchase. Contact Technical Service
Skip To
Iupac Name
ethyl 3-oxo-4-phenylbutanoate
Inchi Code
1S/C12H14O3/c1-2-15-12(14)9-11(13)8-10-6-4-3-5-7-10/h3-7H,2,8-9H2,1H3
InChI key
BOZNWXQZCYZCSH-UHFFFAOYSA-N
storage temp.
Ambient Storage
Purity
90%
Physical Form
Solid
Product Link
https://fluorochem.co.uk/product/F093060
Signal Word
Warning
Hazard Statements
MSDS
https://sds.fluorochem.co.uk/eng/sds_F093060_en.pdf
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
