Sign Into View Organizational & Contract Pricing
Select a Size
Pricing and availability is not currently available. Contact Technical Service
Skip To
Iupac Name
N-(tert-butoxycarbonyl)-2,2-dimethyl-beta-alanine
Inchi Code
1S/C10H19NO4/c1-9(2,3)15-8(14)11-6-10(4,5)7(12)13/h6H2,1-5H3,(H,11,14)(H,12,13)
InChI key
LSZXQEWVQORQBO-UHFFFAOYSA-N
storage temp.
Ambient Storage
Purity
97%
Physical Form
Solid
Product Link
https://fluorochem.co.uk/product/F211010
Signal Word
Warning
Hazard Statements
MSDS
https://sds.fluorochem.co.uk/eng/sds_F211010_en.pdf
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service