Skip to Content
MilliporeSigma

KEY240887155

Key Organics / BIONET

methyl 4-amino-3-bromo-5-methoxybenzoate

Slide 1 of 1
Sign Into View Organizational & Contract Pricing

Select a Size

Pricing and availability is not currently available. Contact Technical Service

About This Item

CAS Number:
Molecular Weight:
260.09
MDL number:

Skip To

Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist

mp

91 - 93

Iupac Name

methyl 4-amino-3-bromo-5-methoxybenzoate

Inchi Code

1S/C9H10BrNO3/c1-13-7-4-5(9(12)14-2)3-6(10)8(7)11/h3-4H,11H2,1-2H3

InChI key

WNTCZYCMVFZROL-UHFFFAOYSA-N

Purity

95%

Country of Origin

GB

Physical Form

Solid

Product Link

https://www.keyorganics.net/sku/LD-0730

Melting Point

91 - 93

pictograms

Exclamation mark

Signal Word

Warning

MSDS

https://www.keyorganics.net/sds/LD-0730


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service