Skip to Content
MilliporeSigma

LEYH9ACFBF2A

Leyan / Shanghai Haohong Scientific Co.,Ltd

Erbium carbonate hydrate, 99.90%

Sign In to View Organizational & Contract Pricing

Select a Size

Pricing and availability is not currently available. Contact Technical Service

Skip To

Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist

Iupac Name

carbonic acid, erbium salt hydrate

Inchi Code

1S/CH2O3.Er.H2O/c2-1(3)4;;/h(H2,2,3,4);;1H2

Inchi Key

NHEBVPYPWOCQOS-UHFFFAOYSA-N

storage temp.

Store at room temperature, keep dry and cool

Purity

99.9%

Country of Origin

CN

Shipping Temperature

room temperature

Physical Form

Solid

Product Link

https://www.leyan.com/22992-83-2.html

pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

MSDS

https://www.leyan.com/22992-83-2.html


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service