Skip to Content
MilliporeSigma

RIE156328385

Rieke Metals, Inc.

3-(3-chloro-5-fluorophenyl)-4'-methylpropiophenone

Sign In to View Organizational & Contract Pricing

Select a Size

1 G

$597.18

2 G

$1,119.70

5 G

$2,239.40

Vendor SKU: 6127-5100-04-1G

$597.18


Ships in 14 days - from Aldrich Partner.


About This Item

CAS Number:
Molecular Weight:
276.74
MDL number:

Skip To

Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist

Iupac Name

3-(3-chloro-5-fluorophenyl)-1-(4-methylphenyl)-1-propanone

Inchi Code

1S/C16H14ClFO/c1-11-2-5-13(6-3-11)16(19)7-4-12-8-14(17)10-15(18)9-12/h2-3,5-6,8-10H,4,7H2,1H3

InChI key

HNBLWYYKDOYTMS-UHFFFAOYSA-N

Purity

97%

Country of Origin

US

Product Link

https://riekemetals.com/products-services/catnum/6127-5100-04

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service