MilliporeSigma
  • IL-33, an interleukin-1-like cytokine that signals via the IL-1 receptor-related protein ST2 and induces T helper type 2-associated cytokines.
IL-33, an interleukin-1-like cytokine that signals via the IL-1 receptor-related protein ST2 and induces T helper type 2-associated cytokines.
Immunity (2005-11-16)
Jochen Schmitz, Alexander Owyang, Elizabeth Oldham, Yaoli Song, Erin Murphy, Terril K McClanahan, Gerard Zurawski, Mehrdad Moshrefi, Jinzhong Qin, Xiaoxia Li, Daniel M Gorman, J Fernando Bazan, Robert A Kastelein
ABSTRACT
Cytokines of the interleukin-1 (IL-1) family, such as IL-1 alpha/beta and IL-18, have important functions in host defense, immune regulation, and inflammation. Insight into their biological functions has led to novel therapeutic approaches to treat human inflammatory diseases. Within the IL-1 family, IL-1 alpha/beta, IL-1Ra, and IL-18 have been matched to their respective receptor complexes and have been shown to have distinct biological functions. The most prominent orphan IL-1 receptor is ST 2. This receptor has been described as a negative regulator of Toll-like receptor-IL-1 receptor signaling, but it also functions as an important effector molecule of T helper type 2 responses. We report a member of the IL-1 family, IL-33, which mediates its biological effects via IL-1 receptor ST 2, activates NF-kappaB and MAP kinases, and drives production of T(H)2-associated cytokines from in vitro polarized T(H)2 cells. In vivo, IL-33 induces the expression of IL-4, IL-5, and IL-13 and leads to severe pathological changes in mucosal organs.
Sigma-Aldrich
IL-33 human, Animal-component free, recombinant, expressed in E. coli, ≥98% (SDS-PAGE), ≥98% (HPLC)
Pricing and availability is not currently available.
Sigma-Aldrich
IL-33 from mouse, recombinant, expressed in E. coli, ≥98% (SDS-PAGE), ≥98% (HPLC), suitable for cell culture
Pricing and availability is not currently available.