MilliporeSigma
Synthesis of Substituted Oxazoles by Visible-Light Photocatalysis.
The Journal of organic chemistry (2016-06-22)
Tanmay Chatterjee, Ji Young Cho, Eun Jin Cho
ABSTRACT
A simple and practical method for the synthesis of substituted oxazoles has been developed using readily available α-bromoketones and benzylamines by visible-light photocatalysis at room temperature. The process, which requires 1 mol % of [Ru(bpy)3]Cl2 photocatalyst with K3PO4 and CCl3Br, is effective for accessing a variety of valuable oxazole compounds. The synthetic utility of our protocol was also demonstrated by preparing a natural product, texaline.
Sigma-Aldrich
[Ru(phen)3]Cl2, ≥95%
Pricing and availability is not currently available.