MilliporeSigma
  • NF-kappaB2 is required for the establishment of central tolerance through an Aire-dependent pathway.
NF-kappaB2 is required for the establishment of central tolerance through an Aire-dependent pathway.
The Journal of clinical investigation (2006-10-14)
Mingzhao Zhu, Robert K Chin, Peter A Christiansen, James C Lo, Xiaojuan Liu, Carl Ware, Ulrich Siebenlist, Yang-Xin Fu
ABSTRACT
NF-kappaB2-deficient mice have impaired T and B cell responses. We found, however, that in these mice there was severe infiltration of lymphocytes into multiple organs and increased activity of autoantibodies to peripheral tissue antigens in a manner similar to that of autoimmune regulator-deficient (Aire-deficient) mice. We further demonstrated that NF-kappaB2 was required for thymic Aire gene transcriptional regulation. The Nfkb2(-/-) thymus had distinct cortical and medullar structures, but reduced Aire and target gene expression of peripheral tissue antigens. Engraftment of Nfkb2(-/-) thymic stroma to nude mice recapitulated the autoimmune phenotype of the native Nfkb2(-/-) mice, confirming a key defect in central tolerance. Lymphotoxin beta receptor (LTbetaR) ligation-induced Aire gene expression was also largely abolished in the absence of NF-kappaB2. Thus NF-kappaB2 downstream of LTbetaR plays an important role in the regulation of central tolerance in an Aire-dependent manner.
Sigma-Aldrich
NFKB2 (1-454), GST tagged human, recombinant, expressed in baculovirus infected Sf9 cells, ≥70% (SDS-PAGE), buffered aqueous glycerol solution
Pricing and availability is not currently available.