MilliporeSigma
  • Fungicide and Cultivar Management of Leaf Spot Diseases of Winter Wheat in Western Canada.
Fungicide and Cultivar Management of Leaf Spot Diseases of Winter Wheat in Western Canada.
Plant disease (2018-08-21)
H R Kutcher, T K Turkington, D L McLaren, R B Irvine, G S Brar
ABSTRACT
A complex of leaf-spotting diseases-tan spot, the Septoria complex, spot blotch, and powdery mildew-are frequently observed on winter wheat in western Canada; however, there are few studies indicating varietal differences in reaction to these diseases or the benefit of fungicide application. To determine the benefit of varietal improvement and multiple fungicide treatments and application timings, field experiments were conducted at six site-years in western Canada. Two cultivars varying in reaction to leaf spot diseases were used in combination with fungicide treatments. Disease severity ranged from trace to 64% of the combined flag and penultimate leaf area diseased, differed between cultivars, and was reduced from the check by some fungicide treatments. Yield improvement by fungicide treatment varied from 3.3 to 13.2% greater than the nontreated check. At two site-years, the split application of two half rates of fungicide resulted in the greatest yield; however, in both cases, the yield benefit did not differ from a single application at the flag leaf growth stage. Cultivar selection and fungicide use under environments conducive to disease are beneficial components of an integrated leaf spot disease management program for winter wheat in western Canada.
Product Description
Supelco
Trifloxystrobin, PESTANAL®, analytical standard
Pricing and availability is not currently available.