MilliporeSigma
  • Encapsulation of monomyristin into polymeric nanoparticles improved its in vitro antiproliferative activity against cervical cancer cells.
Encapsulation of monomyristin into polymeric nanoparticles improved its in vitro antiproliferative activity against cervical cancer cells.
Colloids and surfaces. B, Biointerfaces (2018-12-28)
Sirirat Boondireke, Michèle Léonard, Alain Durand, Benjamas Thanomsub Wongsatayanon
ABSTRACT
The cytotoxicity of monomyristin (MM), a monoacylglycerol, was investigated against cervical cancer cells (HeLa) and two normal cells (Vero and endometrial epithelial cells). MM exhibited cytotoxicity specifically to HeLa cells and not against normal cells except at the highest investigated doses (> 500 μg/mL). MM was showed to increase apoptotic dead cells by intrinsic mitochondrial pathway. To overcome the poor water solubility of MM and increase its efficacy against HeLa cells, MM was encapsulated into dextran-covered polylactide (PLA) nanoparticles (NPs). NPs comprised a PLA core which encapsulated MM and a superficial layer of dextran loops which was used for conjugating a protein, transferrin (Tf), known to be overexpressed on cancer cells' surface. Encapsulation of MM into NPs increased its cytotoxicity against HeLa cells at lower doses of MM than free MM. Additionally, the presence of conjugated Tf further increased the cytotoxicity of MM against HeLa cells as compared to non-conjugated NPs. Remarkably, both conjugated and non-conjugated MM loaded NPs were safe to normal cells (Vero and endometrial).
Sigma-Aldrich
rac-Glycerol 1-myristate, ≥99%
Pricing and availability is not currently available.