MilliporeSigma
  • The nuclear receptor FXR inhibits Glucagon-Like Peptide-1 secretion in response to microbiota-derived Short-Chain Fatty Acids.
The nuclear receptor FXR inhibits Glucagon-Like Peptide-1 secretion in response to microbiota-derived Short-Chain Fatty Acids.
Scientific reports (2020-01-15)
Sarah Ducastel, Véronique Touche, Mohamed-Sami Trabelsi, Alexis Boulinguiez, Laura Butruille, Margaux Nawrot, Simon Peschard, Oscar Chávez-Talavera, Emilie Dorchies, Emmanuelle Vallez, Jean-Sébastien Annicotte, Steve Lancel, Olivier Briand, Kadiombo Bantubungi, Sandrine Caron, Laure B Bindels, Nathalie M Delzenne, Anne Tailleux, Bart Staels, Sophie Lestavel
ABSTRACT
The gut microbiota participates in the control of energy homeostasis partly through fermentation of dietary fibers hence producing short-chain fatty acids (SCFAs), which in turn promote the secretion of the incretin Glucagon-Like Peptide-1 (GLP-1) by binding to the SCFA receptors FFAR2 and FFAR3 on enteroendocrine L-cells. We have previously shown that activation of the nuclear Farnesoid X Receptor (FXR) decreases the L-cell response to glucose. Here, we investigated whether FXR also regulates the SCFA-induced GLP-1 secretion. GLP-1 secretion in response to SCFAs was evaluated ex vivo in murine colonic biopsies and in colonoids of wild-type (WT) and FXR knock-out (KO) mice, in vitro in GLUTag and NCI-H716 L-cells activated with the synthetic FXR agonist GW4064 and in vivo in WT and FXR KO mice after prebiotic supplementation. SCFA-induced GLP-1 secretion was blunted in colonic biopsies from GW4064-treated mice and enhanced in FXR KO colonoids. In vitro FXR activation inhibited GLP-1 secretion in response to SCFAs and FFAR2 synthetic ligands, mainly by decreasing FFAR2 expression and downstream Gαq-signaling. FXR KO mice displayed elevated colonic FFAR2 mRNA levels and increased plasma GLP-1 levels upon local supply of SCFAs with prebiotic supplementation. Our results demonstrate that FXR activation decreases L-cell GLP-1 secretion in response to inulin-derived SCFA by reducing FFAR2 expression and signaling. Inactivation of intestinal FXR using bile acid sequestrants or synthetic antagonists in combination with prebiotic supplementation may be a promising therapeutic approach to boost the incretin axis in type 2 diabetes.
Product Description
Millipore
Glucagon Like Peptide-1 (Active) ELISA, This Glucagon Like Peptide-1 (Active) ELISA is used to measure & quantify Glucagon Like Peptide-1 levels in Metabolism & Endocrine research.
Pricing and availability is not currently available.
Sigma-Aldrich
4-CMTB, ≥98% (HPLC)
Pricing and availability is not currently available.