MilliporeSigma
  • The hemocompatibility of the modified polysulfone membrane with 4-(chloromethyl)benzoic acid and sulfonated hydroxypropyl chitosan.
The hemocompatibility of the modified polysulfone membrane with 4-(chloromethyl)benzoic acid and sulfonated hydroxypropyl chitosan.
Colloids and surfaces. B, Biointerfaces (2020-01-10)
Sheng Yan, Ming-Ming Tu, Yun-Ren Qiu
ABSTRACT
Polysulfone (PSf) membrane is widely employed in blood purification fields, but the blood compatibility of PSf membrane is not adequate. To improve the hemocompatibility of PSf membrane, 4-(chloromethyl)benzoic acid (CMBA) and sulfonated hydroxypropyl chitosan (SHPCS) were grafted onto PSf membrane surface. In our strategy, CMBA was firstly grafted on the PSf membrane surface through the Friedel-Crafts alkylation reaction, and the product was named BAPSf membrane. Then, SHPCS was grafted onto the BAPSf membrane surface by esterification, and the product was named SHPCS-BAPSf membrane. The effects of temperature and reaction time on the productivity of BAPSf and the grafting density of carboxyl and the effects of reaction time on the grafting density of SHPCS grafted onto the BAPSf membrane surface were studied. The SHPCS-BAPSf membranes are investigated by ATR-FTIR, XPS, contact angle measurements and evaluated by blood compatibility in vitro. The results reveal that the hydrophilicity of SHPCS-BAPSf membranes were grealy improved and the evaluation of protein adsorption, hemolysis test, platelet adhesion plasma recalcification time(PRT), activated partial thromboplastin time(APTT), prothrombin time(PT) and thrombin time(TT) confirmed that the SHPCS-BAPSf membranes have remarkable blood compatibility.
Sigma-Aldrich
4-(Chloromethyl)benzoic acid, 95%
Pricing and availability is not currently available.