MilliporeSigma
  • Highly Integrated Elastic Island-Structured Printed Circuit Board with Controlled Young's Modulus for Stretchable Electronics.
Highly Integrated Elastic Island-Structured Printed Circuit Board with Controlled Young's Modulus for Stretchable Electronics.
Micromachines (2020-07-08)
Duho Cho, Junhyung Kim, Pyoenggeun Jeong, Wooyoung Shim, Su Yeon Lee, Youngmin Choi, Sungmook Jung
ABSTRACT
A stretchable printed circuit board (PCB), which is an essential component of next-generation electronic devices, should be highly stretchable even at high levels of integration, as well as durable under repetitive stretching and patternable. Herein, an island-structured stretchable PCB composed of materials with controlled Young's modulus and viscosity by adding a reinforcing agent or controlling the degree of crosslinking is reported. Each material was fabricated with the most effective structures through a 3D printer. The PCB was able to stretch 71.3% even when highly integrated and was patterned so that various components could be mounted. When fully integrated, the stress applied to the mounted components was reduced by 99.9% even when stretched by over 70%. Consequently, a 4 × 4 array of capacitance sensors in a stretchable keypad demonstration using our PCB was shown to work, even at 50% stretching of the PCB.
Sigma-Aldrich
5-Amino-1-pentanol, ≥92.0% (GC)
Pricing and availability is not currently available.
Sigma-Aldrich
Poly(dimethylsiloxane), viscosity 1.0 cSt (25 °C)
Pricing and availability is not currently available.