MilliporeSigma
  • More on target with protein phosphorylation: conferring specificity by location.
More on target with protein phosphorylation: conferring specificity by location.
Trends in biochemical sciences (1996-08-01)
M C Faux, J D Scott
ABSTRACT
One important regulatory mechanism in the control of phosphorylation events is the subcellular location of phosphatases of kinases. Several serine/threonine phosphatases and kinases have now been shown to be associated with targeting subunits; these contribute to the organization and specificity of signal transduction pathways by favoring the accessibility of their enzymes to certain substrates.
Sigma-Aldrich
Protein Phosphatase-1 Catalytic Subunit, α-Isoform from rabbit, 5,000-15,000 units/mg protein, recombinant, expressed in E. coli, lyophilized powder
Pricing and availability is not currently available.