MilliporeSigma
  • Controlling the behaviour of Drosophila melanogaster via smartphone optogenetics.
Controlling the behaviour of Drosophila melanogaster via smartphone optogenetics.
Scientific reports (2020-10-21)
Ilenia Meloni, Divya Sachidanandan, Andreas S Thum, Robert J Kittel, Caroline Murawski
ABSTRACT
Invertebrates such as Drosophila melanogaster have proven to be a valuable model organism for studies of the nervous system. In order to control neuronal activity, optogenetics has evolved as a powerful technique enabling non-invasive stimulation using light. This requires light sources that can deliver patterns of light with high temporal and spatial precision. Currently employed light sources for stimulation of small invertebrates, however, are either limited in spatial resolution or require sophisticated and bulky equipment. In this work, we used smartphone displays for optogenetic control of Drosophila melanogaster. We developed an open-source smartphone app that allows time-dependent display of light patterns and used this to activate and inhibit different neuronal populations in both larvae and adult flies. Characteristic behavioural responses were observed depending on the displayed colour and brightness and in agreement with the activation spectra and light sensitivity of the used channelrhodopsins. By displaying patterns of light, we constrained larval movement and were able to guide larvae on the display. Our method serves as a low-cost high-resolution testbench for optogenetic experiments using small invertebrate species and is particularly appealing to application in neuroscience teaching labs.
Sigma-Aldrich
Anti-Green Fluorescent Protein (GFP) antibody, Mouse monoclonal, clone GFP-20, purified from hybridoma cell culture
Pricing and availability is not currently available.